siRNA Details
| Gene Symbol: Sowahb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_223228.4 | XP_223228.1 | 1 | 2600 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sowahb| Gene Symbol: Sowahb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_223228.4 | XP_223228.1 | 1 | 2600 | ||