siRNA Details
| Gene Symbol: Ccdc158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770088.1 | XP_008768310.1 | 5 | 4921 | ||
| XM_008770089.1 | XP_008768311.1 | 6 | 4189 | |||
| XM_008770090.1 | XP_008768312.1 | 5 | 4909 | |||
| XM_008770091.1 | XP_008768313.1 | 6 | 2102 | |||
| XM_008770092.1 | XP_008768314.1 | 5 | 2032 | |||
| XM_008770093.1 | XP_008768315.1 | 5 | 4921 | |||
| XM_008770094.1 | XP_008768316.1 | 5 | 4921 | |||

Ccdc158
Genomic Map