siRNA Details
| Gene Symbol: Ankrd17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105999.1 | NP_001099469.1 | 2, 3 | 1132 | ||
| XM_006250763.2 | XP_006250825.1 | 2, 3 | 669 | |||
| XM_006250764.2 | XP_006250826.1 | 2, 3 | 669 | |||
| XM_006250765.2 | XP_006250827.1 | 2, 3 | 615 | |||
| XM_006250766.2 | XP_006250828.1 | 2, 3 | 1251 | |||
| XM_006250767.2 | XP_006250829.1 | 2, 3 | 1551 | |||
| XM_006250768.2 | XP_006250830.1 | 2, 3 | 669 | |||
| XM_006250769.2 | XP_006250831.1 | 2, 3 | 669 | |||
| XM_006250770.2 | XP_006250832.1 | 2, 3 | 615 | |||
| XM_008770011.1 | XP_008768233.1 | 2, 3 | 1251 | |||

Ankrd17
Genomic Map