siRNA Details
| Gene Symbol: Enam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106001.1 | NP_001099471.1 | 10 | 1942 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Enam| Gene Symbol: Enam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106001.1 | NP_001099471.1 | 10 | 1942 | ||