siRNA Details
| Gene Symbol: Prr27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250795.2 | XP_006250857.1 | 5 | 897 | ||
| XM_006250796.2 | XP_006250858.1 | 5 | 897 | |||
| XM_006250797.2 | XP_006250859.1 | 4 | 873 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prr27| Gene Symbol: Prr27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250795.2 | XP_006250857.1 | 5 | 897 | ||
| XM_006250796.2 | XP_006250858.1 | 5 | 897 | |||
| XM_006250797.2 | XP_006250859.1 | 4 | 873 | |||