siRNA Details
| Gene Symbol: Tmprss11g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008554.1 | NP_001008554.1 | 7 | 642 | ||
| GenBank mRNA | AJ617481.1 | - | 6 | 603 | ||
| AJ617528.1 | - | 7 | 642 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmprss11g| Gene Symbol: Tmprss11g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008554.1 | NP_001008554.1 | 7 | 642 | ||
| GenBank mRNA | AJ617481.1 | - | 6 | 603 | ||
| AJ617528.1 | - | 7 | 642 | |||