siRNA Details
| Gene Symbol: Noa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006959.1 | NP_001006960.1 | 1 | 263 | ||
| XM_006250845.2 | XP_006250907.1 | 1 | 578 | |||
| GenBank mRNA | BC083902.1 | - | 1 | 263 | 7133605 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Noa1| Gene Symbol: Noa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006959.1 | NP_001006960.1 | 1 | 263 | ||
| XM_006250845.2 | XP_006250907.1 | 1 | 578 | |||
| GenBank mRNA | BC083902.1 | - | 1 | 263 | 7133605 | |