siRNA Details
| Gene Symbol: Fip1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008295.1 | NP_001008296.1 | 7 | 585 | ||
| XM_006250882.2 | XP_006250944.1 | 7 | 689 | |||
| XM_006250883.2 | XP_006250945.1 | 7 | 689 | |||
| XM_006250884.2 | XP_006250946.1 | 7 | 687 | |||
| XM_006250885.2 | XP_006250947.1 | 7 | 687 | |||
| XM_006250886.2 | XP_006250948.1 | 7 | 691 | |||
| XM_006250887.2 | XP_006250949.1 | 7 | 685 | |||
| GenBank mRNA | BC085767.1 | - | 7 | 585 | 7192362 | |
| EU224276.1 | - | 7 | 584 | |||

Fip1l1
Genomic Map