siRNA Details
Gene Symbol: Corin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182473.1 | NP_872279.1 | 3 | 611 | ||
XM_008770139.1 | XP_008768361.1 | 3 | 658 | |||
GenBank mRNA | AY251285.1 | - | 3 | 611 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Corin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182473.1 | NP_872279.1 | 3 | 611 | ||
XM_008770139.1 | XP_008768361.1 | 3 | 658 | |||
GenBank mRNA | AY251285.1 | - | 3 | 611 |