siRNA Details
| Gene Symbol: Commd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106004.2 | NP_001099474.1 | 4, 5 | 545 | ||
| GenBank mRNA | BC159414.1 | - | 4, 5 | 545 | 8368074 | |
| FQ212906.1 | - | 4, 5 | 554 | |||
| FQ221260.1 | - | 4, 5 | 546 | |||

Commd8
Genomic Map