siRNA Details
| Gene Symbol: Gnpda2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106005.1 | NP_001099475.1 | 5, 6 | 721 | ||
| XM_006251036.2 | XP_006251098.1 | 5, 6 | 765 | |||
| GenBank mRNA | BC168866.1 | - | 5, 6 | 708 | 7460205 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnpda2| Gene Symbol: Gnpda2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106005.1 | NP_001099475.1 | 5, 6 | 721 | ||
| XM_006251036.2 | XP_006251098.1 | 5, 6 | 765 | |||
| GenBank mRNA | BC168866.1 | - | 5, 6 | 708 | 7460205 | |