siRNA Details
| Gene Symbol: Atp8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250974.2 | XP_006251036.1 | 15, 16 | 1583 | ||
| XM_006250975.2 | XP_006251037.1 | 15, 16 | 1583 | |||
| XM_006250980.2 | XP_006251042.1 | 11, 12 | 1069 | |||
| XM_008770164.1 | XP_008768386.1 | 15, 16 | 1613 | |||
| XM_008770165.1 | XP_008768387.1 | 12, 13 | 1321 | |||

Atp8a1
Genomic Map