siRNA Details
Gene Symbol: Ube2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106006.1 | NP_001099476.1 | 3, 4 | 432 | ||
XM_006250934.2 | XP_006250996.1 | 3, 4 | 629 | |||
GenBank mRNA | FQ226880.1 | - | 3, 4 | 424 | ||
FQ233155.1 | - | 3, 4 | 380 | |||
FQ234076.1 | - | 3, 4 | 410 |
