siRNA Details
| Gene Symbol: Pgm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106007.1 | NP_001099477.1 | 2 | 275 | ||
| XM_006250996.2 | XP_006251058.1 | 2 | 283 | |||
| XM_006250997.2 | XP_006251059.1 | 3 | 390 | |||
| GenBank mRNA | BC160893.1 | - | 2 | 280 | 7445317 | |

Pgm2
Genomic Map