siRNA Details
| Gene Symbol: Ldb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106009.1 | NP_001099479.1 | 6 | 873 | ||
| XM_008770199.1 | XP_008768421.1 | 6 | 874 | |||
| XM_008770200.1 | XP_008768422.1 | 6 | 874 | |||
| XM_008770201.1 | XP_008768423.1 | 6 | 874 | |||
| XM_008770202.1 | XP_008768424.1 | 6 | 874 | |||
| XM_008770203.1 | XP_008768425.1 | 6 | 874 | |||
| GenBank mRNA | BC168881.1 | - | 6 | 758 | 8363537 | |

Ldb2
Genomic Map