siRNA Details
| Gene Symbol: Lap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011910.1 | NP_001011910.1 | 12 | 1350 | ||
| GenBank mRNA | BC079381.1 | - | 12 | 1350 | 7111053 | |
| FQ218842.1 | - | 8 | 868 | |||
| FQ235301.1 | - | 10 | 1144 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lap3| Gene Symbol: Lap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011910.1 | NP_001011910.1 | 12 | 1350 | ||
| GenBank mRNA | BC079381.1 | - | 12 | 1350 | 7111053 | |
| FQ218842.1 | - | 8 | 868 | |||
| FQ235301.1 | - | 10 | 1144 | |||