siRNA Details
| Gene Symbol: Evc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170439.1 | NP_001163910.1 | 9, 10 | 1382 | ||
| XM_006251090.2 | XP_006251152.1 | 9, 10 | 1416 | |||
| XM_006251091.1 | XP_006251153.1 | 6, 7 | 879 | |||
| Gene Symbol: LOC102553032 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251109.2 | XP_006251171.1 | 1, 2 | 271 | ||

Evc
Genomic Map