siRNA Details
Gene Symbol: Nop14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106013.1 | NP_001099483.1 | 11, 12 | 1741 | ||
XM_006251192.1 | XP_006251254.1 | 11, 12 | 1746 | |||
GenBank mRNA | FQ218277.1 | - | 10, 11 | 1415 |
