siRNA Details
| Gene Symbol: Morc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106016.1 | NP_001099486.1 | 7 | 607 | ||
| XM_006251198.2 | XP_006251260.1 | 9 | 750 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Morc2| Gene Symbol: Morc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106016.1 | NP_001099486.1 | 7 | 607 | ||
| XM_006251198.2 | XP_006251260.1 | 9 | 750 | |||