siRNA Details
| Gene Symbol: Polm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011912.1 | NP_001011912.1 | 10 | 1629 | ||
| XM_006251211.2 | XP_006251273.1 | 10 | 1844 | |||
| XM_006251212.2 | XP_006251274.1 | 10 | 1832 | |||
| XM_006251213.2 | XP_006251275.1 | 10 | 2010 | |||
| XM_006251214.2 | XP_006251276.1 | 10 | 1252 | |||
| GenBank mRNA | BC081868.1 | - | 10 | 1629 | 7096894 | |

Polm
Genomic Map