siRNA Details
Gene Symbol: Cep68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_214106.7 | XP_214106.4 | 3 | 543 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cep68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_214106.7 | XP_214106.4 | 3 | 543 |