siRNA Details
| Gene Symbol: Erlec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106023.1 | NP_001099493.2 | 7 | 660 | ||
| XM_006251615.2 | XP_006251677.1 | 7 | 978 | |||
| GenBank mRNA | FQ228963.1 | - | 6 | 400 | ||
| FQ232613.1 | - | 6 | 399 | |||

Erlec1
Genomic Map