siRNA Details
| Gene Symbol: Efs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106033.2 | NP_001099503.1 | 6 | 1944 | ||
| GenBank mRNA | BC161942.1 | - | 6 | 1944 | 9023909 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Efs| Gene Symbol: Efs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106033.2 | NP_001099503.1 | 6 | 1944 | ||
| GenBank mRNA | BC161942.1 | - | 6 | 1944 | 9023909 | |