siRNA Details
| Gene Symbol: Rec8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011916.1 | NP_001011916.1 | 4 | 434 | ||
| GenBank mRNA | AY529701.1 | - | 4 | 450 | ||
| BC079023.1 | - | 4 | 434 | 7113870 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rec8| Gene Symbol: Rec8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011916.1 | NP_001011916.1 | 4 | 434 | ||
| GenBank mRNA | AY529701.1 | - | 4 | 450 | ||
| BC079023.1 | - | 4 | 434 | 7113870 | ||