siRNA Details
| Gene Symbol: Ipo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106038.1 | NP_001099508.1 | 12 | 1194 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ipo4| Gene Symbol: Ipo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106038.1 | NP_001099508.1 | 12 | 1194 | ||