siRNA Details
| Gene Symbol: Gzmn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191116.1 | NP_001178045.1 | 3 | 300 | ||
| XM_006252044.2 | XP_006252106.2 | 5 | 415 | |||
| XM_008770696.1 | XP_008768918.1 | 4 | 398 | |||
| XM_008770697.1 | XP_008768919.1 | 5 | 466 | |||
| XM_008770698.1 | XP_008768920.1 | 5 | 441 | |||
| XM_008770699.1 | XP_008768921.1 | 4 | 399 | |||
| XM_008770700.1 | XP_008768922.1 | 2 | 183 | |||
| XM_008770701.1 | XP_008768923.1 | 4 | 398 | |||

Gzmn
Genomic Map