siRNA Details
Gene Symbol: Mphosph8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017375.2 | NP_001017375.2 | 9, 10 | 2052 | ||
GenBank mRNA | BC099116.1 | - | 9, 10 | 1995 | 7462184 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mphosph8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017375.2 | NP_001017375.2 | 9, 10 | 2052 | ||
GenBank mRNA | BC099116.1 | - | 9, 10 | 1995 | 7462184 |