siRNA Details
Gene Symbol: Tnfrsf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044229.2 | NP_001037694.1 | 6 | 628 | ||
GenBank mRNA | CO396223.1 | - | 2 | 174 | ||
DQ466087.1 | - | 6 | 628 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tnfrsf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044229.2 | NP_001037694.1 | 6 | 628 | ||
GenBank mRNA | CO396223.1 | - | 2 | 174 | ||
DQ466087.1 | - | 6 | 628 |