siRNA Details
Gene Symbol: Tnfrsf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044229.2 | NP_001037694.1 | 8 | 1124 | ||
GenBank mRNA | AW527025.1 | - | 2 | 258 | ||
CO396223.1 | - | 4 | 670 | |||
DQ466087.1 | - | 8 | 1124 |
