siRNA Details
| Gene Symbol: Ints9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770801.1 | XP_008769023.1 | 12, 13 | 1540 | ||
| Gene Symbol: LOC102549712 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252224.2 | XP_006252286.1 | 1151 | |||

Ints9
Genomic Map