siRNA Details
| Gene Symbol: Loxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106047.2 | NP_001099517.2 | 13 | 2417 | ||
| GenBank mRNA | BC168900.1 | - | 13 | 2417 | 9032601 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Loxl2| Gene Symbol: Loxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106047.2 | NP_001099517.2 | 13 | 2417 | ||
| GenBank mRNA | BC168900.1 | - | 13 | 2417 | 9032601 | |