siRNA Details
| Gene Symbol: Klhl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106054.1 | NP_001099524.1 | 4 | 1648 | ||
| XM_008770864.1 | XP_008769086.1 | 4 | 1804 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl1| Gene Symbol: Klhl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106054.1 | NP_001099524.1 | 4 | 1648 | ||
| XM_008770864.1 | XP_008769086.1 | 4 | 1804 | |||