siRNA Details
| Gene Symbol: Mycbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106055.2 | NP_001099525.2 | 31, 32 | 5474 | ||
| XM_008770938.1 | XP_008769160.1 | 4235 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mycbp2| Gene Symbol: Mycbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106055.2 | NP_001099525.2 | 31, 32 | 5474 | ||
| XM_008770938.1 | XP_008769160.1 | 4235 | ||||