siRNA Details
| Gene Symbol: Cldn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106058.1 | NP_001099528.1 | 3, 4 | 786 | ||
| XM_006252450.2 | XP_006252512.1 | 2, 3 | 627 | |||
| XM_006252451.2 | XP_006252513.1 | 2, 3 | 519 | |||
| GenBank mRNA | BC158829.1 | - | 2, 3 | 389 | ||

Cldn10
Genomic Map