siRNA Details
| Gene Symbol: Uggt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252469.2 | XP_006252531.1 | 3 | 860 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uggt2| Gene Symbol: Uggt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252469.2 | XP_006252531.1 | 3 | 860 | ||