siRNA Details
| Gene Symbol: Anxa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011918.1 | NP_001011918.1 | 7 | 886 | ||
| XM_008770980.1 | XP_008769202.1 | 7 | 1132 | |||
| GenBank mRNA | BC083812.1 | - | 7 | 886 | 7110191 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Anxa11| Gene Symbol: Anxa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011918.1 | NP_001011918.1 | 7 | 886 | ||
| XM_008770980.1 | XP_008769202.1 | 7 | 1132 | |||
| GenBank mRNA | BC083812.1 | - | 7 | 886 | 7110191 | |