siRNA Details
| Gene Symbol: Nt5dc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009271.1 | NP_001009271.1 | 10, 11 | 1156 | ||
| XM_008770986.1 | XP_008769208.1 | 10, 11 | 3198 | |||
| GenBank mRNA | AY569011.1 | - | 10, 11 | 1149 | ||
| BC088431.1 | - | 10, 11 | 1156 | 7124640 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_027289, BC011230.

Nt5dc2
Genomic Map