siRNA Details
| Gene Symbol: Nt5dc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009271.1 | NP_001009271.1 | 6 | 721 | ||
| XM_008770986.1 | XP_008769208.1 | 6 | 2763 | |||
| GenBank mRNA | AY569011.1 | - | 6 | 714 | ||
| BC088431.1 | - | 6 | 721 | 7124640 | ||

Nt5dc2
Genomic Map