siRNA Details
| Gene Symbol: Stab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771053.1 | XP_008769275.1 | 50 | 5552 | ||
| XM_008771054.1 | XP_008769276.1 | 50 | 5552 | |||
| XM_008771055.1 | XP_008769277.1 | 51 | 5384 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stab1| Gene Symbol: Stab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771053.1 | XP_008769275.1 | 50 | 5552 | ||
| XM_008771054.1 | XP_008769276.1 | 50 | 5552 | |||
| XM_008771055.1 | XP_008769277.1 | 51 | 5384 | |||