siRNA Details
| Gene Symbol: Ogdhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106062.1 | NP_001099532.1 | 10, 11 | 1469 | ||
| XM_006252584.2 | XP_006252646.1 | 1664 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ogdhl| Gene Symbol: Ogdhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106062.1 | NP_001099532.1 | 10, 11 | 1469 | ||
| XM_006252584.2 | XP_006252646.1 | 1664 | ||||