siRNA Details
| Gene Symbol: Chat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170593.1 | NP_001164064.1 | 1707 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chat| Gene Symbol: Chat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170593.1 | NP_001164064.1 | 1707 | |||