siRNA Details
| Gene Symbol: Wdfy4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252759.2 | XP_006252821.1 | 30, 31 | 5379 | ||
| XM_006252760.2 | XP_006252822.1 | 30, 31 | 6920 | |||
| XM_006252762.2 | XP_006252824.1 | 30, 31 | 5380 | |||
| XM_008771159.1 | XP_008769381.1 | 31, 32 | 5463 | |||

Wdfy4
Genomic Map