siRNA Details
| Gene Symbol: LOC290595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106063.2 | NP_001099533.2 | 3 | 237 | ||
| GenBank mRNA | AF152002.1 | - | 3 | 240 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC290595| Gene Symbol: LOC290595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106063.2 | NP_001099533.2 | 3 | 237 | ||
| GenBank mRNA | AF152002.1 | - | 3 | 240 | ||