siRNA Details
| Gene Symbol: Ghitm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005908.1 | NP_001005908.1 | 8 | 1074 | ||
| XM_006252786.2 | XP_006252848.1 | 8 | 1328 | |||
| GenBank mRNA | BC083778.1 | - | 8 | 1074 | 7193683 | |
| FQ212260.1 | - | 8 | 1274 | |||
| FQ217853.1 | - | 8 | 1092 | |||

Ghitm
Genomic Map