siRNA Details
| Gene Symbol: RGD1311847 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013879.1 | NP_001013901.1 | 2 | 199 | ||
| XM_006252799.2 | XP_006252861.1 | 2 | 256 | |||
| XM_008771073.1 | XP_008769295.1 | 2 | 249 | |||
| GenBank mRNA | BC079430.1 | - | 2 | 199 | 7114707 | |

RGD1311847
Genomic Map