siRNA Details
| Gene Symbol: Colgalt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106067.2 | NP_001099537.1 | 10 | 1275 | ||
| GenBank mRNA | BC160899.1 | - | 10 | 1275 | 7453718 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Colgalt1| Gene Symbol: Colgalt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106067.2 | NP_001099537.1 | 10 | 1275 | ||
| GenBank mRNA | BC160899.1 | - | 10 | 1275 | 7453718 | |