siRNA Details
Gene Symbol: Ifi30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030026.1 | NP_001025197.1 | 2, 3 | 445 | ||
GenBank mRNA | BC088256.1 | - | 2, 3 | 309 | ||
BC099774.1 | - | 2, 3 | 445 | 7932222 | ||
FQ225096.1 | - | 2, 3 | 331 | |||
FQ229307.1 | - | 2, 3 | 483 | |||
FQ230297.1 | - | 2, 3 | 331 | |||
FQ232607.1 | - | 2, 3 | 331 | |||
FQ233058.1 | - | 2, 3 | 331 | |||
FQ233063.1 | - | 2, 3 | 363 | |||
FQ233214.1 | - | 2, 3 | 331 | |||
FQ234315.1 | - | 2, 3 | 331 |
