siRNA Details
| Gene Symbol: Tktl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106080.1 | NP_001099550.1 | 1 | 784 | ||
| XM_006253022.2 | XP_006253084.1 | 1 | 809 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tktl2| Gene Symbol: Tktl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106080.1 | NP_001099550.1 | 1 | 784 | ||
| XM_006253022.2 | XP_006253084.1 | 1 | 809 | |||