siRNA Details
Gene Symbol: Cep44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037181.1 | NP_001032258.1 | 4 | 521 | ||
XM_006253087.2 | XP_006253149.1 | 3 | 350 | |||
XM_006253089.2 | XP_006253151.1 | 4 | 585 | |||
XM_008771223.1 | XP_008769445.1 | 4 | 780 | |||
GenBank mRNA | BC107470.1 | - | 4 | 521 | 7457548 | |
FQ228732.1 | - | 4 | 585 |
