siRNA Details
Gene Symbol: Dctd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013882.2 | NP_001013904.2 | 71 | |||
GenBank mRNA | BC087138.1 | - | 26 | 7125992 | ||
DY317307.1 | - | 39 | ||||
EV776012.1 | - | 96 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dctd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013882.2 | NP_001013904.2 | 71 | |||
GenBank mRNA | BC087138.1 | - | 26 | 7125992 | ||
DY317307.1 | - | 39 | ||||
EV776012.1 | - | 96 |